About this item
The Mouse Dendritic Cell backbone panel is a six-color combination of anti-mouse antibodies. This panel can serve as a backbone for identifying major dendritic cell (DC) subsets. Hosts: Rat, Armenian Hamster
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7, M1/70, N418, RA3-6B2, RB6-8C5, M5/114.15.2
Conjugation: PE (Phycoerythrin)-Cy7®, APC (Allophycocyanin)-Cy7, PerCP-Cy5.5, APC (Allophycocyanin), PE (Phycoerythrin), FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Multiple
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10002-352
- 10002-354
- Antigen symbol:
- CD8A
- CD8A
- Antigen name:
- CD8 antigen, alpha chain
- CD8 antigen, alpha chain
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 53-6.7, M1/70, N418, RA3-6B2, RB6-8C5, M5/114.15.2
- 53-6.7, M1/70, N418, RA3-6B2, RB6-8C5, M5/114.15.2
- Reactivity:
- Mouse
- Mouse
- Host:
- Multiple
- Multiple