About this item
The Mouse Myeloid Precursor backbone panel is a five-color combination of seven anti-mouse antibodies. This panel can serve as a backbone to identify various myeloid precursor populations, and leaves room for GFP or additional drop-in markers, if desired. The CD3e, CD19 and CD45R/B220 positive cells are gated together to exclude mature T and B cells. Violet laser (405 nm) required for detection of CD11b violetFluorTM 450. Hosts: Rat, Armenian Hamster
Contains:
1 x redFluor™ 710 Anti-Mouse CD3 (17A2)
1 x violetFluor™ 450 Anti-Human/Mouse CD11b (M1/70)
1 x PerCP-Cyanine5.5 Anti-Mouse CD11c (N418)
1 x redFluor™ 710 Anti-Mouse CD19 (1D3)
1 x redFluor™ 710 Anti-Human/Mouse CD45R (B220) (RA3-6B2)
1 x PE-Cyanine7 Anti-Mouse Ly-6G (Gr-1) (RB6-8C5)
1 x FITC Anti-Mouse TER-119 (TER-119)
Type: Primary
Antigen: CD3, CD11b, CD11c, CD19, CD45R, Ly-6G, TER-119
Clonality: Monoclonal
Clone: 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-8C5, TER-119
Conjugation: redFluor® 710, violetFluor® 450, PerCP-Cy5.5, PE-Cy7, FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Multiple
Isotype:
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10002-350
- 10002-348
- Antigen symbol:
- CD3
- CD3
- Antibody type:
- Primary
- Primary
- Conjugation:
- Multiple
- Multiple
- Clone:
- 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-8C5, TER-119
- 17A2, M1/70, N418, 1D3, RA3-6B2, RB6-8C5, TER-119
- Reactivity:
- Mouse
- Mouse
- Host:
- Multiple
- Multiple
- Flow cytometry:
- Yes
- Yes