About this item
The MY31 antibody reacts with human CD56, also known as the neural cell adhesion molecule (NCAM), a glycoprotein which is a member of the immunoglobulin superfamily. The 140 kDa isoform of CD56 is expressed on human NK cells and NK-T cells, with increased expression levels on activated NK lymphocytes. The CD56 antigen is also expressed by neurons and is reported to play a role in nervous system development and neural cell-to-cell adhesion.
Type: Primary
Antigen: NCAM1
Clonality: Monoclonal
Clone: MY31
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10002-322
- 10002-320
- Antigen name:
- Neural Cell Adhesion Molecule 1
- Neural Cell Adhesion Molecule 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- MY31
- MY31
- Host:
- Mouse
- Mouse
- Gene ID:
- 4684
- 4684
- Isotype:
- IgG1 kappa
- IgG1 kappa