About this item
The PC61.5 antibody is specific for mouse CD25, a 55 kDa surface protein also known as the Interleukin-2 Receptor alpha chain, or IL-2R alpha. CD25 may bind IL-2 by itself, although with low affinity and without induction of cell signaling. CD25 is also expressed within a high-affinity complex, along with the IL-2R beta chain (CD122) and the common gamma chain (CD132), to form a signaling receptor complex. Expression of CD25 varies during developmental stages of T and B cells, is induced on activated mature T and B cells, and is present on subsets of dendritic cells. CD25 signaling as part of the IL-2 receptor complex triggers T cell activation and proliferation, as well as modulating the differentiation and function of Th17 cells, T regulatory (Treg) cells, and dendritic cells. The PC61.5 antibody is used as a marker for T cells, B cells and dendritic cell subsets. Expression of CD25, CD4 and the transcription factor Foxp3 is regarded as a phenotypic signature for Treg cells. As such, this antibody is widely used to distinguish Treg cells from na�ve or conventional T cells which are CD25-. This clone has also been reported for depletion of Treg cells in vivo (use format suitable for functional assays).
Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: PC61.5
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG1 lambda
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10002-192
- 10002-190
- 10002-194
- Antigen name:
- Interleukin 2 Receptor, Alpha
- Interleukin 2 Receptor, Alpha
- Interleukin 2 Receptor, Alpha
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- PC61.5
- PC61.5
- PC61.5
- Host:
- Rat
- Rat
- Rat
- Gene ID:
- 16184
- 16184
- 16184
- Isotype:
- IgG1 kappa
- IgG1 lambda
- IgG1 kappa