About this item
The 2H7 antibody reacts with human CD20, expressed by pre-B cells, resting and activated B cells, follicular dendritic cells and a small subset of T cells, but not on plasma cells. CD20 is a 33-37 kDa transmembrane protein which is differentially phosphorylated in resting and active B cells. The CD20 molecule plays a role in regulating B-cell activation.
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 2H7
Conjugation: violetFluor® 450
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10753-836
- 10753-840
- 10753-838
- Antigen name:
- Membrane-Spanning 4-Domains, Subfamily A, Member 1
- Membrane-Spanning 4-Domains, Subfamily A, Member 1
- Membrane-Spanning 4-Domains, Subfamily A, Member 1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- 2H7
- 2H7
- 2H7
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 931
- 931
- 931
- Isotype:
- IgG2b kappa
- IgG2b kappa
- IgG2b kappa
Specifications
Related Information
tds-75-0209.pdf
Datasheet
Tonbo Biosciences violetFluor™ Antibodies.pdf