About this item
This inexpensive set of materials outfits your whole class for gel electrophoresis at a great price.
- Precursor to working with DNA and proteins
- Material for six lab groups
- Engaging activity for students
- Estimated lab time: 45 minutes
- Inexpensive system set
These electrophoresis dyes can be used as a precursor to working with DNA and proteins. When the colorful dyes are electrophoresed, they separate by their molecular size and charge. Students compare the electrophoresed results of the individual dyes to the dye mixture to find the missing component.
Demo Size requires 5 alkaline or rechargeable 9v batteries
Class Size requires 30 alkaline or rechargeable 9v batteries