Aluminum magnesium (alloy 5056) (95:5 %) (w/w), gauze woven from wire 20 mesh Ø 0.23 mm (0.009 in)
Supplier: Avantor
Certificates
About this item
Formula:Al:Mg;95:5wt%
MDL Number:MFCD00214039
:Aluminum magnesium (alloy 5056)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:20 mesh Ø 0.23 mm (0.009 in)
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Aluminum magnesium (alloy 5056) (95:5 %) (w/w), gauze woven from wire 20 mesh Ø 0.23 mm (0.009 in):
Specification Test Results
Related Information
Certificates
Search by Lot NumberAlfa Aesar Battery Technology 2021.pdf Alfa Aesar Metals and Materials
Product Literature
Alfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
Customers Who Bought This Also Bought