Filter Vial Press and Toggle Press for Thomson SINGLE StEP Standard Filter Vials, Restek
Supplier: Restek
Certificates
About this item
Accessories for Thomson SINGLE StEP Standard Filter Vials.
- Filter vial press with multiple uses
- Compatible with 30 mL filter vials and autosampler-ready filter vials
The filter vial press boasts 8 positions for 30 mL filter vials, and 48 positions for autosampler-ready filter vials.
Customers Who Bought This Also Bought