Glutathione Sepharose™ 4B Affinity Chromatography Media, Cytiva
Supplier: Cytiva
Certificates
About this item
Glutathione Sepharose™ 4B is designed for high capacity single-step purification of Glutathione S-Transferase (GST) tagged fusion proteins.
- Proteins are eluted under mild, non-denaturing conditions that preserve protein antigenicity and functionality
Related Information
Certificates
Search by Lot Number17-0756-01 42 BR 17-0756-01 42 BR (1) 17-0756-01 42 LIT 17-0756-01 42 LIT (3) 17-0756-01_42 DF (1).pdf
Datasheet
More
17-0756-01_42 IN.pdf 17-0756-04_42 IN.pdf 17-0756-05_42 IN.pdf 17075601_HAND BOOK 1.pdf 17075601_HAND BOOK 2.pdf
Manual
More
95016-984_10299550
Conformity declaration
Customers Who Bought This Also Bought