Calmodulin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Catalog #: CA95016-874L
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:90 µm
- pH range:4 - 9
- Description:Calmodulin Sepharose™ 4B
- Flow rate:<75 cm/h
- Size:10 mL
- Matrix:Agarose, 4%
- Ligand:Calmodulin
- Storage temperature:2 - 8 °C
- Cat. no.:CA95016-874L
- Type:Liquid
- Supplier No.:17-0529-01
Specifications
About this item
Calmodulin Sepharose™ 4B is calmodulin immobilised by the CNBr method to Sepharose™ 4B. Calmodulin is a highly conserved regulatory protein found in all eukaryotic cells. This protein is involved in many cellular processes such as glycogen metabolism, cytoskeletal control, neurotransmission, phosphate activity and control of NAD⁺/NADP⁺.
- For single-step purification of native calmodulin-binding proteins
- Suitable for tandem affinity purification (TAP) of protein complexes
- Purification of calmodulin-regulated proteins from all eukaryotic cells
- Cyanogen bromide activation coupling method
- Stable to all commonly used aqueous solutions
- Negligible volume variation due to changes in pH or ionic strength
Calmodulin binds proteins principally through their interactions with hydrophobic sites on its surface. These sites are exposed after a conformational change induced by the action of Ca²⁺ on separate Ca²⁺ binding sites. The binding of enzymes may be enhanced if the enzyme substrate is present and enzymesubstratecalmoldulin-Ca²⁺ complexes are particularly stable.
Customers Who Bought This Also Bought