Equilibration Tubes, Cytiva
Catalog #: CA11028-370
Supplier: Cytiva
Specifications
- Description:Equilibration Tubes
- Cat. no.:CA11028-370
- Supplier No.:80646779
Specifications
About this item
For equilibrating IPG strips prior to second dimension.
Ordering information: Includes recommendation regarding equilibration solution volumes.
Customers Who Bought This Also Bought