Inlet Filter Set, Cytiva
Catalog #: CA10498-002
Supplier: Cytiva
Specifications
- Description:Inlet Filter Set, Polypropylene
- Cat. no.:CA10498-002
- Supplier No.:18-1114-42
Specifications
About this item
Set contents include 10 net rings and 10 support screens
The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, with the exception of 100% Ethyl acetate, 100% Hexane, and 100% Tetrahydrofuran.
Customers Who Bought This Also Bought