Inlet Filter Assembly Kit, Cytiva
Catalog #: CA10497-990
Supplier: Cytiva
Specifications
- Description:Inlet Filter Assembly Kit, Polypropylene and Titanium Alloy
- Cat. no.:CA10497-990
- Supplier No.:18-1113-15
Specifications
About this item
Kit contents include one support net, one filter (mounted on fliterholder), and one screw lid and cap membrane.
The wetted parts, except for the membrane, are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, with the exception of 100% Ethyl acetate, 100% Hexane, and 100% Tetrahydrofuran.
Customers Who Bought This Also Bought