Gelatin Sepharose™ 4B Affinity Chromatography Media, Cytiva
Catalog #: CA95017-042L
Supplier: Cytiva
CAS Number:
Specifications
- Particle size:90 µm
- pH range:3 - 10
- Description:Gelatin Sepharose™ 4B
- Flow rate:<75 cm/h
- Size:25 mL
- Matrix:Agarose, 4%
- Ligand:Bovine gelatin
- Storage temperature:2 - 8 °C
- Ligand density:4.5-8 mg gelatin/ml drained resin
- Ligand coupling method:Cyanogen bromide activation
- Cat. no.:CA95017-042L
- Type:Liquid
- Supplier No.:17-0956-01
Specifications
About this item
Gelatin Sepharose® 4B affinity media with immobilised gelatin is designed for the purification or depletion of fibronectin as gelatin binds to fibronectin with high specificity. Gelatin Sepharose® 4B is bovine gelatin coupled to Sepharose® 4B by the cyanogen bromide method.
Gelatin binds specifically to fibronectin, which is a high molecular weight glycoprotein found on the surfaces of many cell types and present in many extracellular fluids including plasma. Gelatin Sepharose 4B has been designed for the purification or removal of fibronectin.
Customers Who Bought This Also Bought