Whatman™ Grade 593½ Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Cytiva
Certificates
About this item
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. A standard grade filter paper for fine precipitates.
- Increased filter surface area increases total loading capacity and decreases overall filtration time
- Reduced contact between filter and funnel maintains flow rate
- Eliminated need to quadrant-fold filter circles to fit conical funnels saves time in repetitive or multiple analyses
- Prepleated filters have the same technical properties as the respective filter circles
Prepleated qualitative filters give improved flow rate and increased loading capacity compared to equivalent quadrant folded filters.
Specifications
- Catalog No:
- CA11012-582
- CA11012-568
- Grade:
- Grade 593
- Grade 593
- Thickness:
- 170 µm (typical)
- 170 µm (typical)
- Particle retention:
- 5 µm (in liquid max.)
- 5 µm (in liquid max.)
- Filtration speed:
- 450 (Herzberg)
- 450 (Herzberg)
- Basis weight:
- 85 g/m²
- 85 g/m²
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Customers Who Bought This Also Bought