Digital Depth Gage Assembly, Quality Lab Accessories
Catalog #: 89132-992
Supplier: QLA
Specifications
- Description:Digital Depth Gauge Assembly
- Cat. no.:89132-992
- Supplier No.:DDAPT-Q
Specifications
About this item
Digital Depth Gage Assembly for accurate measurement of Dissolution Paddle and Basket height within the dissolution vessel.
Delivery: Includes all required components and storage case.
Customers Who Bought This Also Bought