To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:ATP-binding cassette, Subfamily C (CFTR/MRP), Member 10
- Clonality:Polyclonal
- Gene ID:89845
- Host:Multiple
- Reactivity:Human
- Antigen symbol:ABCC10
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Antigen synonyms:SIMRP7|EST182763|MRP7|ATP binding cassette subfamily C member 10
- Cat. no.:10535-754
- Supplier No.:H00089845-PW1
Specifications
About this item
This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.
Antibody pair set content: 1. Antibody pair for IP: Rabbit Polyclonal Anti-ABCC10 (300 µl); 2. Antibody pair for Western Blot: Mouse purified Polyclonal Anti-ABCC10 (50 µg)
Type:
Antigen: ABCC10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human