To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:Angiotensin II Receptor-Associated Protein
- Gene ID:57085
- Host:Multiple
- Reactivity:Human
- Antigen symbol:AGTRAP
- Antigen synonyms:Angiotensin Γàí receptor associated protein|ATI receptor associated protein|ATRAP_HUMAN.|Angiotensin Γàí type I receptor associated protein|Type 1 angiotensin Γàí receptor associated protein|ATRAP|MGC29646
- Cat. no.:10535-952
- Supplier No.:H00057085-AP11
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human AGTRAP.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-AGTRAP (100 µg); 2. Detection Antibody: Mouse Monoclonal Anti-AGTRAP, IgG1 kappa (20 µg);
Type:
Antigen: AGTRAP
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human