To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antigen name:BAI1-associated protein 2
- Gene ID:10458
- Host:Multiple
- Reactivity:Human
- Antigen symbol:BAIAP2
- Antigen synonyms:FLAF3|brain-specific angiogenesis inhibitor 1-associated protein 2|fas ligand-associated factor 3|BAIAP2|BAP2|BAI1-associated protein 2|insulin receptor substrate p53|IRSP53|insulin receptor substrate p53/p58
- Cat. no.:10536-104
- Supplier No.:H00010458-AP51
Specifications
About this item
This Set comes with matched Antibody Pair to detect and quantify protein level of human BAIAP2.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-BAIAP2 (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-BAIAP2 (50 µg);
Type:
Antigen: BAIAP2
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human