FlexTech™ Gloves, White Poly/Cotton Shell with Latex Palm, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
These palm-coated work gloves provide additional protection in high-wear or risk areas of the hand.
- White polyester/cotton string knit shell with blue latex palm coating
- Latex coating offers superior grip and abrasion and puncture resistance
- Excellent for canning, recycling and general manufacturing
- 10 Gauge
The Y9243 have a polyester/cotton string knit shell with a blue latex palm coating. The coated glove shells are excellent for a variety of applications that require a high level of dexterity along with cut protection.
Non-slip surface allows workers to work more efficiently and with greater confidence. The open, uncoated back circulates air, allowing hands to ‘breathe’ and keeping them cool and comfortable.
Ideal for use in construction.
Certifications: ANSI Cut Level 2.
Ordering information: Gloves are sold by the dozen pairs.
Specifications
- Catalog No:
- 80092-360
- 80092-358
- 80092-356
- 80092-354
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Polyester/cotton
- Finish:
- Latex palm coating
- Latex palm coating
- Latex palm coating
- Latex palm coating
- Gauge:
- 10
- 10
- 10
- 10
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist