To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Oxford Listeria Selective Supplement
- Size:10 x 1 vial
- Solubility:(20 °C) soluble
- Incubation:48 hrs. at 35 °C; aerobic
- Storage:Store at +2°C to +8°C.
- Cat. no.:CA1.07006.0010
- Supplier No.:1.07006.0010
Specifications
About this item
Supplement tested in Oxford Listeria selective agar (base) art
- Greater than or equal to 70% recovery on test medium Listeria monocytogenes ATCC 19111 (WDCM 00020)
- Greater than or equal to 70% recovery on test medium Listeria monocytogenes ATCC 13932 (WDCM 00021)
- Greater than or equal to 70% recovery on test medium Listeria monocytogenes ATCC 35152 (WDCM 00109)
- Greater than or equal to 70% recovery on test medium Listeria innocua ATCC 33090 (WDCM 00017)
- Less than or equal to 0.01% recovery on test medium Enterococcus faecalis ATCC 19433 (WDCM 00009)
- Less than or equal to 0.01% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
- Black zone Listeria monocytogenes ATCC 19111 (WDCM 00020) +
- Black zone Listeria monocytogenes ATCC 13932 (WDCM 00021) +
- Black zone Listeria monocytogenes ATCC 35152 (WDCM 00109) +
- Black zone Listeria innocua ATCC 33090 (WDCM 00017) +
- Growth inhibited in Staphylococcus aureus ATCC 25923 (WDCM 00034)
- Growth inhibited in Candida albicans ATCC 10231 (WDCM 00054)
- Reference Tryptic soy agar
In North America 1.07006.0010 replaces and is identical to 1.07006.0017
no. 1.07004. Cycloheximide ( >= 90 % - <= 100 % ) 66-81-9 and acriflavine hydrochloride ( >= 1 % - < 5 % ) 69235-50-3.