Pierce™ Water Soluble Siliconizing Fluid
Catalog #: 10066-324
Supplier: Thermo Scientific
Specifications
- Description:AquaSil siliconizing fluid
- Size:120 ml
- Supplier No.:TS-42799
- Chemical Compound:4-hydroxy-4-methylpentan-2-one, 2-methyl-2-propanol, silane, trimethoxyoctadecyl
Specifications
About this item
Ideal for siliconizing glass surfaces.
- Greater resistance to base hydrolysis than other surface treatments
- Can be used on plastic surfaces
It is easy-to-use silane monomer solution is supplied as a 20% solid solution in a mixture of diacetone alcohol and tertiary butyl alcohol. The primary silane component is an octadecyltrialkoxysilane, which is hydrolyzed to a silanol when mixed with water.
Hazards: Flammable, irritant
Standard components: 4-hydroxy-4-methylpentan-2-one, 2-methyl-2-propanol, silane, trimethoxyoctadecyl
Standard concentration: 25 to 45, 25 to 45, 10 to 20%
Storage: 15 to 35 °C
Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought