Resprep® SPE Flow Filters, Restek
Catalog #: 10059-484
Supplier: Restek
Specifications
- Description:Resprep® SPE Flow Filters
- Cat. no.:10059-484
- Supplier No.:26024
Specifications
About this item
These SPE Flow Filters are designed to be used with Resprep® Diskcover-47 reservoir, or any 47 mm glass sample reservoir.
- Improved flow when filtering oil and grease samples
Customers Who Bought This Also Bought