Shelving for CApture™ BT Fuming Chamber, Labconco
Supplier: Labconco
Certificates
About this item
Wire and perforated shelf kits for evidence to sit on or hang from for fuming in the CApture™ BT Fuming Chamber.
- All shelves are made of stainless steel
- Weight limit of wire shelves: 25 lbs. per shelf
- Weight limit of half shelves: 15 lbs. each
- Weight limit of perforated shelves: 25 lbs. per shelf
The increased solid surface on these shelves provides greater stability and makes them ideal for small evidence.
Each shelf fits onto CApture™ BT sidewall brackets for easy installation and removal. The CApture™ BT can hold up to 4 full-size wire shelves, 8 half-depth shelves, or 3 perforated shelves.
Delivery: The half shelf kit is available which includes two wire half-depth shelves.
Customers Who Bought This Also Bought