EDU Culture Dish Control System, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
Certificates
About this item
The EDU Culture Dish Control System is a simplified version of the Delta T® system
- Enhance the instruction of biology
- Includes curriculum relevant demonstrations
- Temperature selection options
- Basic X, Y translation built in
The EDU Starter Set includes all of the necessary components required for full system operation. The EDU Controller and Power Supply is an electronic, closed-loop temperature controller interfaced to EDU stage adapters.
It is designed to facilitate demonstrations of live cellular events in a classroom setting.
Certifications: CE certified.
Ordering information: EDU Starter Set includes, EDU controller, power supply, EDU stage adapter, one pack of 10 dishes, one hundred 22 mm coverslips, 22 mm glass spacer, coverslip weight, and 6 mm culture cylinder. When purchased individually, the controller and power supply comes with a multi-voltage AC input, 12 VDC output power supply. Also available separately, the EDU Stage Adapter is a metal plate containing a circuit board, temperature sensor, and electrical contacts that accept Delta T® dishes and enable X,Y translation of the specimen on any microscope.
Customers Who Bought This Also Bought