Stable "Z"® Heated Lid and Controller, Bioptechs Inc.®
Catalog #: 10199-848
Supplier: BIOPTECHS INC.
Specifications
- Description:Stable "Z"® Heated Lid and Controller
- Cat. no.:10199-848
- Supplier No.:40319260812
Specifications
About this item
Designed for use with the Stable "Z" system to prevent condensation and provide a more uniform temperature gradient across the dish of the Stable "Z".
- Prevents condensation
- Prevents outside contamination
Certifications: CE certified.
Delivery: Includes lid, controller, and power supply.
Customers Who Bought This Also Bought