Stable "Z"® Starter Set, Bioptechs Inc.®
Catalog #: 10199-838
Supplier: BIOPTECHS INC.
Specifications
- Description:Stable "Z"® Starter Set
- Cat. no.:10199-838
- Supplier No.:4031926
Specifications
About this item
The Stable "Z"® is a device designed to warm biological specimens in standard plasticware on a microscope stage, with the distinct advantage of eliminating "Z" axis drift.
- Minimal mass of heating component
- Specimen is coplanar with support surface to eliminate "Z" axis drift
- Minimizes heat transfer to the stage
- Has option of heated lid to prevent condensation
- Adjustable specimen plane to accommodate various dishes
- Option for Nunc Lab-Tec multi-well slides
- Includes a reference temperature probe
Certifications: CE certified.
Ordering information: Set includes controller, power supply, and specified stage adapter.
Customers Who Bought This Also Bought