Objective Cooling Collar, Bioptechs Inc.®
Catalog #: 10199-814
Supplier: BIOPTECHS INC.
Specifications
- Description:Objective Cooling Collar
- Cat. no.:10199-814
- Supplier No.:150303
Specifications
About this item
Ring-shaped attachment to an objective provides an isolated pathway through which a refrigerated fluid can flow, to provide an efficient means of cooling the objective.
Certifications: CE certified.
Customers Who Bought This Also Bought