Objective Wrench and Plier Set, Bioptechs
Catalog #: 10199-798
Supplier: BIOPTECHS INC.
Specifications
- Description:Objective Wrench and Plier Set
- Cat. no.:10199-798
- Supplier No.:021523
Specifications
About this item
This tool set used to isolate the mechanical stress applied to an objective when removing the decorative collar.
- Safely allows removal of decorative collar
- For use with the Objective Heater® System
Designed to enable the attachment of an objective heater.
Certifications: CE certified.
Customers Who Bought This Also Bought