¹/₁₆" Perfusion Pump Tubing Adapter Set, Bioptechs Inc.®
Catalog #: 10200-108
Supplier: BIOPTECHS INC.
Specifications
- Description:1/16" Perfusion Pump Tubing Adapter Set
- Cat. no.:10200-108
- Supplier No.:1620031
Specifications
About this item
A kit of 1/16" tubing barbs and connectors that enable fluid connection to FCS2® or FCS3® chambers
Each set includes: two Tee Adapters, two Male Luer Taper Adapters to 1/16" Tubing, two Female 1/16" Tubing to Luer Adapters, and one Two-way Valve.
Certifications: CE certified.
Customers Who Bought This Also Bought