FCS2® Cooling Adapter, Bioptechs Inc.®
Catalog #: 10200-020
Supplier: BIOPTECHS INC.
Specifications
- Description:FCS2® Cooling Adapter
- Cat. no.:10200-020
- Supplier No.:06031920301
Specifications
About this item
The FCS2® Cooling Adapter is an O-Ring sealed window that circulates an isolated cooled fluid over the upper glass element (Microaqueduct slide) to cool the specimen to desired temperature
This Cooling Adapter is designed for use with FCS2® Closed Chamber Systems.
Certifications: CE certified.
Customers Who Bought This Also Bought