Specifications
- Description:Microaqueduct Slides
- Cat. no.:10200-058
- Supplier No.:1301195
Specifications
About this item
A 40mm diameter, 1mm thick glass circle with an ITO coating on the outer surface
Suitable for no flow through high rate flow procedures where a rapid exchange of media is required with low cell surface shear
Cell temperature can be controlled from ambient to 50°C (±0.2°C) without the need of an air curtain
Temperature is controlled uniformly across entire field with media equilibrating as it enters the chamber
Closed system so that bicarbonate CO₂ or organic buffers can be employed
Compatible with ¹⁄₁₆"" tubing for perfusion (C-Flex, Tygon, etc.)
Easily assembled (no tools required)
Stand-alone temperature controller with an alarm circuit to safeguard cells
Near laminar flow
Slides feature two port holes and microaqueduct grooves to facilitate laminar flow perfusion in a Bioptechs FCS2® or FCS3® chambers.