D-(-)-Sorbitol 91.0-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol,(Sorbitol),D(-)-Glucitol
CAS Number:50-70-4
:98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MW:182.17 g/mol
MDL Number:MFCD00004708
:Length:
:Width:
:Height:
:Ø
:Thickness:
:91,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 94 to 98 °C
:Flash Pt:
:Flash Pt:
:M.W. 182.17 g/mol
:D-(-)-Sorbitol
:295
:M.O.C. Category R
:12,08873
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets N.F. Requirements:
- GMP Manufactured Product:
- Meets J.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay (anhydrous basis):91.0 - 100.5 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - pH of 10% solution:3.5 - 7.0
- NF - Reducing Sugars:≤ 0.3 %
- NF - Residue on Ignition:≤ 0.1 %
- NF - Nickel (Ni):≤ 1 ppm
- NF - Chloride (Cl):≤ 0.0050 %
- NF - Sulfate (SO₄):≤ 0.01 %
- NF - Water:≤ 1.5 %
- NF - Total Aerobic Microbial Count, cfu/g:≤ 1000
- NF - Total Yeast and Mold Count, cfu/g:≤ 100
- EP/BP - Assay (anhydrous basis):97.0 - 102.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Conductivity, uS cm-1:≤ 20
- EP/BP - Reducing Sugars:≤ 0.2 %
- EP/BP - Related Substances - Any impurity, each:≤ 2.0 %
- EP/BP - Related Substances - Total:≤ 3.0 %
- EP/BP - Water:≤ 1.5 %
- Total Viable Aerobic Count - Total Aerobic Microbial Count, cfu/g:≤ 100
- Total Viable Aerobic Count - Total Yeast and Mold Count, cfu/g:≤ 100
- Total Viable Aerobic Count - Escherichia coli (negative):Passes Test
- Salmonella (negative):Passes Test
- Endotoxin Concentration, <2.5 IU/g:Passes Test
- JP - Assay (dried basis):≥ 97.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Identification C:Passes Test
- JP - Clarity and Color:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Chloride (Cl):≤ 0.005 %
- JP - Sulfate (SO₄):≤ 0.006 %
- JP - Heavy Metals (as Pb):≤ 5 ppm
- JP - Nickel (Ni):Passes Test
- JP - Arsenic (As):≤ 1.3 ppm
- JP - Glucose:Passes Test
- JP - Sugars:Passes Test
- JP - Loss on Drying:≤ 2.0 %
- JP - Residue on Ignition:≤ 0.02 %
- Only Class 2 solvents (Ethylene Glycol) are likely to be present. All are below Option 1 limits.:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
sds_BAKR3449
SDS
spec_JT3449-06 spec_JT3449-9
Product specification
Customers Who Bought This Also Bought