About this item
The CApture™ BT is a spacious benchtop Cyanoacrylate (CA) evidence fuming chamber with a durable, easy-to-clean stainless steel liner
The microprocessor-controlled operating system allows for adjustment of chamber humidity to 80% +/- 5% relative humidity, pre-fuming humidity incubation, fuming temperature up to 424°F within +/- 1%, and fuming and exhaust times. System stores 20 programs, has 3 factory set programs (Low, High, Lumincyano) and includes diagnostic mode with audible/visual alerts for program cycle completion. Acti-Vent™ Smart Controls purge air during critical steps to protect evidence from over fuming exposure and the user from fumes.
The CApture™ BT is constructed of a powder-coated steel exterior, type 304 brushed stainless steel interior with removable perforated floor, four stainless steel hanging rods (26.9" long x 3/8" dia.) and quick-release sidewall brackets mounted at four heights. A storage drawer (28Wx13Dx1.5H") with flush-mounted handle is located beneath the chamber.
It features a built-in humidifier, full-color eye-level LCD control screen, recirculation blower, exhaust blower and interior LED light. The tempered safety glass door has an electronic door lock to prevent opening during the fuming cycle and includes a power outage override. Filters are accessible from inside the chamber for easy replacement.
Base stand adds portability and storage to CApture™ BT Fuming Chamber. Powder-coated steel frame with 5" diameter, toe-locking, polyurethane casters. Features shelves and compartment to hold supplies and accessories. Stand height is manually adjustable from 21" to 28". Weight capacity of the stand is 400 lbs."
Its ample interior (19 cubic feet with 58.6" diagonal length) with shelving system maximizes fuming capacity and accommodates large evidence including up to 15 long guns.
Super Glue, fuming dishes, and purified water are required (not included).
Specifications
- Catalog No:
- 10014-196
- 10014-198
- 10014-200
- 10014-336
- 10014-338
- 10014-340
- 10014-346
- Environmentally Preferable:
- Sustainable Materials, Energy Efficient
- Sustainable Materials, Energy Efficient
- Sustainable Materials, Energy Efficient
- Sustainable Materials, Energy Efficient
- Sustainable Materials, Energy Efficient
- Sustainable Materials, Energy Efficient