Replacement Light Bulb, Labconco
Catalog #: 10769-374
Supplier: Labconco
Product available on GSA Advantage®
Specifications
- Description:Replacement Light Bulb, 9x2x1"
- Cat. no.:10769-374
- Supplier No.:9577700
Specifications
About this item
Pre-wired vapor-proof fluorescent lighting provides up to 63 foot candles
Lights are located within the hood above the work surface for increased visibility.
Ordering information: For use with Protector® Filtered Fume Hoods. Quantity required per model; 3' Hood - 1 light; 4' Hood - 2 lights; 5' Hood - 3 lights; 6' Hood - 4 lights; 8' Hood - 5 lights. First set of bulbs (of number required based on hood width) included with each Protector Echo Filtered Fume Hood.
Customers Who Bought This Also Bought