Anti-YPYDVPDYA Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog #: 10065-800
Supplier: Columbia Biosciences
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Tag
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:YPYDVPDYA
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Storage temperature:Store at 4 °C
- Tested applications:High Throughput Screening, Fluorescence Resonance Energy Transfer Assays, Flow Cytometry and Microscopy
- Cat. no.:10065-800
- Supplier No.:D3-1830
Specifications
About this item
Type: Tag
Antigen: YPYDVPDYA
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Customers Who Bought This Also Bought