Anti-LRRC26 Rabbit Polyclonal Antibody
Catalog #: 89520-190
Supplier: Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Leucine Rich Repeat Containing 26
- Clonality:Polyclonal
- Gene ID:389816
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:LRRC26
- Conjugation:Unconjugated
- ELISA:Yes
- Size:400 µL
- Western blot:Yes
- Epitope:C-Terminal
- Antigen synonyms:BK channel auxiliary gamma subunit LRRC26|LRRC26|bA350O14.10|CAPC|Leucine-rich repeat-containing protein 26
- Amino acid number:289 to 318
- Storage buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular weight:34.857 kDa
- Sequence:PPRPPDPNPDPDPHGC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping temperature:4 °C
- Immunogen:This LRRC26 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 289-318 amino acids from the C-terminal region of human LRRC26.
- Purification:Protein A & Peptide affinity purification
- Cat. no.:89520-190
- Supplier No.:AP16312B
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: LRRC26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human
Customers Who Bought This Also Bought