To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD45R
- Clonality:Monoclonal
- Clone:RA3-6B2
- Gene ID:19264, 5788
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse,Cat
- Antigen symbol:CD45R
- Conjugation:Brilliant Violet® 570
- Flow cytometry:Yes
- Size:125 µL
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and BSA (origin USA).
- Antigen synonyms:B220|PTPRC
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Immunogen:Abelson murine leukemia virus-induced pre-B tumor cells
- Purification:The antibody was purified by affinity chromatography and conjugated with Brilliant Violet 570™ under optimal conditions. The solution is free of unconjugated Brilliant Violet 570™ and unconjugated antibody.
- Cat. no.:CA103237-BL
- Supplier No.:103237
Specifications
About this item
Brilliant Violet 570™ anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cat (Feline); Apps: FC; Size: 125 μl
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: Brilliant Violet® 570
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cat (Feline)