To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:LT-alpha
- Clonality:Monoclonal
- Clone:359-81-11
- Gel shift:Yes
- Gene ID:4049
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen symbol:LT-alpha
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Size:500 µg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Antigen synonyms:Natural killer cytotoxic factor (NKCF)|Hemorrhagic factor|Lymphotoxin-α (LT- α)|TNFSF-1|Coley’s toxin|Differentiation inducing factor (DIF)|TNFB|Necrosin|Tumor necrosis factor-β
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C. Do not freeze.
- Concentration:0.50 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:<em>E. coli</em> expressed, recombinant human LT-α.
- Purification:The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
- Cat. no.:CA503104-BL
- Supplier No.:503104
Specifications
About this item
Biotin anti-human LT-α (TNF-β) [359-81-11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Detection, ELISPOT Detection, ICFC; Size: 500 μg
Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.25-1.0 μg/ml is recommended. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: LT-alpha
Clonality: Monoclonal
Clone: 359-81-11
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human