To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD8
- Clonality:Monoclonal
- Clone:SK1
- Gene ID:925
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:African green monkey,Cynomolgus monkey,Sooty mangabey,Rhesus monkey,Pigtailed macaque,Chimpanzee,Human
- Antigen symbol:CD8
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Size:100 Tests
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Antigen synonyms:T8|Leu2
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 488 under optimal conditions.
- Cat. no.:CA344716-BL
- Supplier No.:344716
Specifications
About this item
Alexa Fluor® 488 anti-human CD8 [SK1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: African Green, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus, Sooty Mangabey; Apps: FC, IHC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: SK1
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: African Green;Chimpanzee;Cynomolgus;Pigtailed Macaque;Rhesus;Sooty Mangabey