About this item
Purified anti-human IL-21 [3A3-N2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: ELISA Capture; Size: 200 μg
Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-21 recombinant protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: IL-21
Clonality: Monoclonal
Clone: 3A3-N2
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Rhesus
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA513002-BL
- CA513001-BL
- Antigen name:
- IL-21
- IL-21
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 3A3-N2
- 3A3-N2
- Host:
- Mouse
- Mouse
- Gene ID:
- 59067
- 59067
- Gel shift:
- Yes
- Yes