About this item
PE/Cy5 anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cat (Feline); Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD45R
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cat (Feline)
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA103209-BL
- CA103210-BL
- Antigen name:
- CD45R
- CD45R
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- RA3-6B2
- RA3-6B2
- Host:
- Rat
- Rat
- Gene ID:
- 19264, 5788
- 19264, 5788
- Isotype:
- IgG2a kappa
- IgG2a kappa