About this item
Biotin anti-mouse/human NGF [JKhNGF-2]; Isotype: Mouse IgG1; Reactivity: Mouse, Human, 2.5S, 7S, and β-NGF forms Cross-Reactivity: Cattle (Bovine, Cow), Viper; Apps: ELISA Detection; Size: 500 μg
Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 1 - 3 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application. Additionally, BioLegend recommends the usage of Tris-Buffered Saline (TBS), pH = 7.2 for all washes and incubations, except substrate buffer. Do not use PBS as results may vary dramatically.
Type: Primary
Antigen: NGF
Clonality: Monoclonal
Clone: JKhNGF-2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Mouse;Human;2.5S;7S;and β-NGF forms Cross-Reactivity: Cattle (Bovine;Cow);Viper
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA509801-BL
- CA509802-BL
- Antigen name:
- NGF
- NGF
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- JKhNGF-2
- JKhNGF-2
- Host:
- Mouse
- Mouse
- Gene ID:
- 18049, 4803
- 18049, 4803
- Gel shift:
- Yes
- Yes