To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD120b (TNF R Type II/p75)
- Clonality:Monoclonal
- Clone:TR75-89
- Gene ID:21938
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Antigen symbol:CD120b (TNF R Type II/p75)
- Conjugation:Biotin
- Flow cytometry:Yes
- Size:50 µg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Antigen synonyms:TNFR1A|TNFRSF1B|p75|TNFR type II|TNFRII|TNFR80
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C. Do not freeze.
- Concentration:0.50 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:<I>E. coli</I> -expressed mouse Type II TNFR
- Purification:The antibody was purified by affinity chromatography, and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
- Cat. no.:CA113403-BL
- Supplier No.:113403
Specifications
About this item
Biotin anti-mouse CD120b (TNF R Type II/p75) [TR75-89]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 50 μg
Each lot of this antibody is quality control tested by ELISA assay and/or immunofluorescent staining with flow cytometric analysis For ELISA detection applications, a concentration range of 0.125-0.5 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of TNF R Type II/p75 recombinant protein ranging from 250 to 4 pg/ml are recommended for each ELISA plate. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD120b (TNF R Type II/p75)
Clonality: Monoclonal
Clone: TR75-89
Conjugation: Biotin
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse