To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD195 (CCR5)
- Clonality:Monoclonal
- Clone:HEK/1/85a
- Gene ID:1234
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Human
- Antigen symbol:CD195 (CCR5)
- Conjugation:Alexa Fluor® 700
- Flow cytometry:Yes
- Size:25 µg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Antigen synonyms:CCR5|C-C chemokine receptor type 5|HIV-1 fusion co-receptor
- Storage temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Concentration:0.50 mg/mL
- Shipping temperature:Ambient RT
- Immunogen:CHO cells transfected with human CCR5
- Purification:The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 700 under optimal conditions.
- Cat. no.:CA313713-BL
- Supplier No.:313713
Specifications
About this item
Alexa Fluor® 700 anti-human CD195 (CCR5) [HEK/1/85a]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is highly recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD195 (CCR5)
Clonality: Monoclonal
Clone: HEK/1/85a
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human