About this item
Alexa Fluor® 647 anti-human CD195 (CCR5) [HEK/1/85a]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD195 (CCR5)
Clonality: Monoclonal
Clone: HEK/1/85a
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA313711-BL
- CA313712-BL
- Antigen name:
- CD195 (CCR5)
- CD195 (CCR5)
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HEK/1/85a
- HEK/1/85a
- Host:
- Rat
- Rat
- Gene ID:
- 1234
- 1234
- Isotype:
- IgG2a kappa
- IgG2a kappa