Anti-RAD51 Mouse Monoclonal Antibody [clone: 51RAD01]
Supplier: Diagnostic Biosystems
Certificates
About this item
Type:
Antigen: RAD51
Clonality: Monoclonal
Clone: 51RAD01
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: 0
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA95024-808
- CA99990-606
- Antigen symbol:
- RAD51
- RAD51
- Antigen name:
- RAD51 recombinase
- RAD51 recombinase
- Antigen synonyms:
- hRAD51|RAD51L3|DNA repair protein RAD51 homolog 1|BRCA1/BRCA2-containing complex|HRAD51|HsRad51|RecA|recombination protein A|HsRAD51|RAD51|HsT16930|subunit 5|RAD51A|E. coli|homolog of|RAD51 homolog A|RAD51D|BRCC5|RECA|RecA-like protein
- hRAD51|RAD51L3|DNA repair protein RAD51 homolog 1|BRCA1/BRCA2-containing complex|HRAD51|HsRad51|RecA|recombination protein A|HsRAD51|RAD51|HsT16930|subunit 5|RAD51A|E. coli|homolog of|RAD51 homolog A|RAD51D|BRCC5|RECA|RecA-like protein
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 51RAD01
- 51RAD01
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
Specifications
Customers Who Bought This Also Bought