Specifications
- Description:Determine the Amino Acid Sequence Kit
- No. of Lab Groups:6
- Cat. no.:470144-308
- Includes:Teacher ‘s Guide and Student Copymasters
- Supplier No.:8-160
Specifications
About this item
This kit determines the amino acid sequence of a tripeptide isolated from a protein hydrolysate.
- Students Determine the Chain Length of Peptide by Formol Titration
- Amino Acid-Focused Kit
- Activities Demonstrate how Chromatographic Techniques Determine Rf Values
- Required Materials Included
With this kit, students determine the chain length of the peptide by formol titration and identify the amino acid content and composition of the tripeptide by using chromatographic techniques to determine Rf values.
Students also identify the N-terminal and C-terminal residues using solvent extraction, enzymatic digestion, and chromatographic techniques.