To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:melanoma cell adhesion molecule
- Clonality:Monoclonal
- Clone:P1H12
- Gene ID:Q8R2Y2
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Antigen symbol:Mcam
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Size:25 tests
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Mcam
- Storage buffer:Phosphate-buffered aqueous solution, ≤0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Cat. no.:10765-802
- Supplier No.:76-886
Specifications
About this item
The P1H12 monoclonal antibody reacts with CD146, 118kDA transmembrane glycoprotein that is also called the melanoma cell adhesion molecule (Mel-CAM), MUC18, Endo-CAM, and S-Endo. It is a prominently glycosylated protein with carbohydrates comprising the majority of its mass and it is expressed on melanoma, stromal, endothelial, and bone marrow fibroblasts. The P1H12 antibody cross reacts with human, mouse, canine, rabbit to the exclusion of rat protein.
Type: Primary
Antigen: Mcam
Clonality: Monoclonal
Clone: P1H12
Conjugation: PE
Epitope:
Host: Mouse
Isotype: Mouse IgG1, kappa
Reactivity: Mouse