To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:histocompatibility 2, K1, K region
- Clonality:Monoclonal
- Clone:34-1-2S
- Gene ID:P01901
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Antigen symbol:H2-K1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Size:0.1 mg
- Cross adsorption:No
- Format:purified antibody
- Form:liquid
- Antigen synonyms:H2-K1
- Storage buffer:Phosphate-buffered aqueous solution, ph7.2.
- Storage temperature:The product should be stored undiluted at 4 °C . Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography. The endotoxin level is determined by LAL test to be less than 0.01 EU/µg of the protein.
- Cat. no.:10765-864
- Supplier No.:77-186
Specifications
About this item
The 34-1-2S monoclonal antibody specifically reacts with the mouse H-2k/H2D MHC class I alloantigens, which are involved in antigen presentation to T cells. The 34-1-2S antibody cross-reacts with the b, p, q, r, and s haplotypes.
Type: Primary
Antigen: H2-K1
Clonality: Monoclonal
Clone: 34-1-2S
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, kappa
Reactivity: Mouse